accessibility menu, dialog, popup

UserName

ClClOPOOClClOClCl

CAS RN 13674-87-8

IUPAC Name: tris(1,3-dichloropropan-2-yl) phosphate
Synonyms: tris(1,3-dichloropropan-2-yl) phosphate
Molecular Weight (g/mol): 430.89
Molecular Formula: C9H15Cl6O4P
InChi Key: ASLWPAWFJZFCKF-UHFFFAOYSA-N
SMILES: ClCC(CCl)OP(=O)(OC(CCl)CCl)OC(CCl)CCl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results