accessibility menu, dialog, popup

UserName

HN

CAS RN 13672-18-9

IUPAC Name: N-benzylnaphthalen-2-amine
Synonyms: N-benzylnaphthalen-2-amine
Molecular Weight (g/mol): 233.31
Molecular Formula: C17H15N
InChi Key: KBOITYDYBXNTMF-UHFFFAOYSA-N
SMILES: C(NC1=CC=C2C=CC=CC2=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results