missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-2-naphthylamine 98.0+%, TCI America™

Catalog No. B30885G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30885G 5 g
B308825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30885G Supplier TCI America Supplier No. B30885G
Only null left

Chemical Identifiers

CAS 13672-18-9
Molecular Formula C17H15N
Molecular Weight (g/mol) 233.314
MDL Number MFCD00144848
InChI Key KBOITYDYBXNTMF-UHFFFAOYSA-N
PubChem CID 12487301
IUPAC Name N-benzylnaphthalen-2-amine
SMILES C1=CC=C(C=C1)CNC2=CC3=CC=CC=C3C=C2

Specifications

Melting Point 68°C
Boiling Point 190°C
Quantity 5 g
Formula Weight 233.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzyl-2-naphthylamine