CAS RN 136496-72-5
IUPAC Name:
2-chloro-4-(dihydroxyboranyl)benzoic acid
Synonyms:
2-chloro-4-(dihydroxyboranyl)benzoic acid
Molecular Weight (g/mol):
200.38
Molecular Formula:
C7H6BClO4
InChi Key:
QFAFGWXQNDYXPZ-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC(Cl)=C(C=C1)C(O)=O