missing translation for 'onlineSavingsMsg'
Learn More

4-Carboxy-3-chlorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C25231G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C25231G Supplier TCI America Supplier No. C25231G
Only null left

Specifications

Chemical Name or Material 4-Carboxy-3-chlorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 136496-72-5
Melting Point 232°C
Color White
Molecular Formula C7H6BClO4
Quantity 1 g
MDL Number MFCD06656268
Synonym 4-carboxy-3-chlorophenylboronic acid, 3-chloro-4-carboxyphenylboronic acid, 4-carboxy-3-chlorobenzeneboronic acid, 2-chloro-4-dihydroxyboranyl benzoic acid, 4-carboxy-3-chlorobenzeneboronicacid, pubchem5134, 4-carboxy-3, akos brn-0544, acmc-1cuk9, ksc174k1d
InChI Key QFAFGWXQNDYXPZ-UHFFFAOYSA-N
SMILES B(C1=CC(=C(C=C1)C(=O)O)Cl)(O)O
IUPAC Name 4-borono-2-chlorobenzoic acid
Molecular Weight (g/mol) 200.381
PubChem CID 14903223
Formula Weight 200.38
Physical Form Crystalline Powder
Show More Show Less