accessibility menu, dialog, popup

UserName

CAS RN 13638-82-9

IUPAC Name: 1,3,6,8-tetraphenylpyrene
Synonyms: 1,3,6,8-tetraphenylpyrene
Molecular Weight (g/mol): 506.65
Molecular Formula: C40H26
InChi Key: SIJHJHYRYHIWFW-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=CC(C2=CC=CC=C2)=C2C=CC3=C(C=C(C4=CC=CC=C4)C4=CC=C1C2=C34)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)