accessibility menu, dialog, popup

UserName

CaH3CCH3OOH3CCH3OO

CAS RN 136-51-6

IUPAC Name: calcium bis(2-ethylhexanoate)
Synonyms: calcium bis(2-ethylhexanoate)
Molecular Weight (g/mol): 326.49
Molecular Formula: C16H30CaO4
InChi Key: LTPCXXMGKDQPAO-UHFFFAOYNA-L
SMILES: [Ca++].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Quantity 
  • (1)
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)