missing translation for 'onlineSavingsMsg'
Learn More

Calcium 2-ethylhexanoate, 98%

Catalog No. AAA1826622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1826622 100 g
AAA1826606 5 g
AAA1826614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1826622 Supplier Thermo Scientific Chemicals Supplier No. A1826622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 136-51-6 | C16H30CaO4 | 326.49 g/mol

Calcium 2-ethylhexanoate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium 2-ethylhexanoate is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 136-51-6
Molecular Formula C16H30CaO4
Molecular Weight (g/mol) 326.49
MDL Number MFCD00014001
InChI Key LTPCXXMGKDQPAO-UHFFFAOYSA-L
Synonym calcium 2-ethylhexanoate, calcium bis 2-ethylhexanoate, hexanoic acid, 2-ethyl-, calcium salt, hexanoic acid, 2-ethyl-, calcium salt 2:1, calcium 2-ethyl-hexanoate, octope ca, calcium hex-cem, nikka octhix calcium, calcium 2-ethylhexoate, dsstox_cid_7063
PubChem CID 61082
IUPAC Name calcium;2-ethylhexanoate
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Ca+2]

Specifications

Odor Odorless
Quantity 100 g
Solubility Information Insoluble in water.
Formula Weight 326.5
Percent Purity 98%
Chemical Name or Material Calcium 2-ethylhexanoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-249-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only