accessibility menu, dialog, popup

UserName

CAS RN 1355326-35-0

H3CONHNFONNH3C

CAS RN 1355326-35-0

IUPAC Name: N-cyclohexyl-2-[N-(3-fluorophenyl)-2-(2-methyl-1H-imidazol-1-yl)acetamido]-2-(2-methylphenyl)acetamide
Synonyms: N-cyclohexyl-2-[N-(3-fluorophenyl)-2-(2-methylimidazol-1-yl)acetamido]-2-(2-methylphenyl)acetamide
Molecular Weight (g/mol): 462.57
Molecular Formula: C27H31FN4O2
InChi Key: FNYGWXSATBUBER-UHFFFAOYNA-N
SMILES: CC1=NC=CN1CC(=O)N(C(C(=O)NC1CCCCC1)C1=CC=CC=C1C)C1=CC(F)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results