accessibility menu, dialog, popup

UserName

CAS RN 13529-27-6

H3COOCH3O

CAS RN 13529-27-6

IUPAC Name: 2-(diethoxymethyl)furan
Synonyms: 2-(diethoxymethyl)furan 2-furaldehyde-diethylacetal
Molecular Weight (g/mol): 170.21
Molecular Formula: C9H14O3
InChi Key: SEILDMUKBMYIEZ-UHFFFAOYSA-N
SMILES: CCOC(OCC)C1=CC=CO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results