missing translation for 'onlineSavingsMsg'
Learn More

2-Furaldehyde Diethyl Acetal 98.0+%, TCI America™

Catalog No. F09295G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09295G 5 g
F092925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09295G Supplier TCI America Supplier No. F09295G
Only null left

Chemical Identifiers

CAS 13529-27-6
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.21
MDL Number MFCD00003245
InChI Key SEILDMUKBMYIEZ-UHFFFAOYSA-N
Synonym 2-diethoxymethyl furan, 2-furaldehyde diethyl acetal, furfural diethyl acetal, furan, 2-diethoxymethyl, unii-04b3gme3ij, 2-furaldehydediethylacetal, 2-diethoxymethyl-furan, furan, diethoxymethyl, 2-furaldehyde diethylacetal, 04b3gme3ij
PubChem CID 83553
ChEBI CHEBI:34769
IUPAC Name 2-(diethoxymethyl)furan
SMILES CCOC(OCC)C1=CC=CO1

Specifications

Color Yellow
Boiling Point 81°C
Quantity 5 g
Formula Weight 170.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Furaldehyde Diethyl Acetal
RTECSNumber LU0585000
TSCA Yes