CAS RN 134356-73-3
IUPAC Name:
2-(4-fluorophenyl)oxirane
Synonyms:
2-(4-fluorophenyl)oxirane
Molecular Weight (g/mol):
138.14
Molecular Formula:
C8H7FO
InChi Key:
ICVNPQMUUHPPOK-UHFFFAOYNA-N
SMILES:
FC1=CC=C(C=C1)C1CO1
1
–
1
of
1
results
1
Promotions Available
(R)-(-)-4-Fluorostyrene oxide, 98+%, ee 98+%, Thermo Scientific Chemicals
(R)-(-)-4-Fluorostyrene oxide, 98+%, ee 98+%, Thermo Scientific Chemicals
CAS: 134356-73-3 Molecular Formula: C8H7FO Molecular Weight (g/mol): 138.14 MDL Number: MFCD03788740,MFCD00043994,MFCD03788740 InChI Key: ICVNPQMUUHPPOK-UHFFFAOYNA-N Synonym: r-4-fluorophenyl oxirane,r-2-4-fluorophenyl oxirane,2r-2-4-fluorophenyl oxirane,r-4-fluorostyrene oxide,r---4-fluorostyrene oxide,pubchem18166,r-4-fluorophenyl oxirane, technical gc PubChem CID: 7023123 IUPAC Name: (2R)-2-(4-fluorophenyl)oxirane SMILES: FC1=CC=C(C=C1)C1CO1