accessibility menu, dialog, popup

UserName

H3CSH

CAS RN 13373-97-2

IUPAC Name: icosane-1-thiol
Synonyms: icosane-1-thiol
Molecular Weight (g/mol): 314.62
Molecular Formula: C20H42S
InChi Key: YYHYWOPDNMFEAV-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCS
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)