missing translation for 'onlineSavingsMsg'
Learn More

1-Eicosanethiol 95.0+%, TCI America™

Catalog No. E08325G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E08325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E08325G Supplier TCI America Supplier No. E08325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13373-97-2
Molecular Formula C20H42S
Molecular Weight (g/mol) 314.616
MDL Number MFCD07779406
InChI Key YYHYWOPDNMFEAV-UHFFFAOYSA-N
Synonym 1-eicosanethiol, eicosanethiol, 1-icosanethiol, icosyl mercaptan, eicosyl mercaptan, acmc-209btf
PubChem CID 139443
IUPAC Name icosane-1-thiol
SMILES CCCCCCCCCCCCCCCCCCCCS

Specifications

Melting Point 40°C
Color White
Boiling Point 389°C
Quantity 5 g
Formula Weight 314.62
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Eicosanethiol
TSCA No
Recommended Storage Refrigerator