missing translation for 'onlineSavingsMsg'
Learn More

1-Eicosanethiol, 98%

Catalog No. AAH34498MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34498MD 250 mg
AAH3449803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH34498MD Supplier Thermo Scientific Chemicals Supplier No. H34498MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13373-97-2 | C20H42S | 314.616 g/mol

1-Eicosanethiol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Eicosanethiol is used as an organic chemical synthesis intermediate.

Notes
Air and moisture sensitive. Incompatible with heat.

Chemical Identifiers

CAS 13373-97-2
Molecular Formula C20H42S
Molecular Weight (g/mol) 314.616
MDL Number MFCD07779406
InChI Key YYHYWOPDNMFEAV-UHFFFAOYSA-N
Synonym 1-eicosanethiol, eicosanethiol, 1-icosanethiol, icosyl mercaptan, eicosyl mercaptan, acmc-209btf
PubChem CID 139443
IUPAC Name icosane-1-thiol
SMILES CCCCCCCCCCCCCCCCCCCCS

Specifications

Melting Point 38°C to 41°C
Odor Stench
Linear Formula CH3(CH2)19SH
Quantity 250 mg
Sensitivity Air sensitive
Formula Weight 314.61
Percent Purity 98%
Chemical Name or Material 1-Eicosanethiol
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only