accessibility menu, dialog, popup

UserName

CAS RN 13322-19-5

CH3NNNHO

CAS RN 13322-19-5

IUPAC Name: (5-methyl-2-phenyl-2H-1,2,3-triazol-4-yl)methanol
Synonyms: (5-methyl-2-phenyl-1,2,3-triazol-4-yl)methanol
Molecular Weight (g/mol): 189.22
Molecular Formula: C10H11N3O
InChi Key: VYHJVMFMRIGUFV-UHFFFAOYSA-N
SMILES: CC1=NN(N=C1CO)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results