missing translation for 'onlineSavingsMsg'
Learn More

(5-Methyl-2-phenyl-2H-1,2,3-triazol-4-yl)methanol, 95%, Thermo Scientific™

Catalog No. CC16209DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC16209DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC16209DA Supplier Thermo Scientific Chemicals Supplier No. CC16209DA
Only null left

Chemical Identifiers

CAS 13322-19-5
Molecular Formula C10H11N3O
Molecular Weight (g/mol) 189.22
MDL Number MFCD02682048
InChI Key VYHJVMFMRIGUFV-UHFFFAOYSA-N
Synonym 5-methyl-2-phenyl-2h-1,2,3-triazol-4-yl methanol, 2h-1,2,3-triazole-4-methanol, 5-methyl-2-phenyl, 5-methyl-2-phenyl-1,2,3-triazol-4-yl methanol, 5-methyl-2-phenyl-1,2,3-triazol-4-yl methan-1-ol, 2-phenyl-4-hydroxymethyl-5-methyl-2h-1,2,3-triazole, 5-methyl-2-phenyltriazol-4-yl methanol, 2h-1,2,3-triazole-4-methanol,5-methyl-2-phenyl, 5-methyl-2-phenyl-1,2,3-triazol4-yl methan-1-ol, 5-methyl-2-phenyl-2h-1,2,3 triazol-4-yl-methanol, 5-methyl-2-phenyl-2h-1,2,3-triazol-4-yl methanol #
PubChem CID 563195
IUPAC Name (5-methyl-2-phenyl-2H-1,2,3-triazol-4-yl)methanol
SMILES CC1=NN(N=C1CO)C1=CC=CC=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 189.22
Percent Purity 95%
Chemical Name or Material (5-Methyl-2-phenyl-2H-1, 2, 3-triazol-4-yl)methanol
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.