accessibility menu, dialog, popup

UserName

CH3H3CCH3OONOHONNN

CAS RN 132622-65-2

IUPAC Name: 4-azido-1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid
Synonyms: 4-azido-1-(tert-butoxycarbonyl)pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 256.26
Molecular Formula: C10H16N4O4
InChi Key: JZEOWBJXFSZTJU-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CC(CC1C(O)=O)N=[N+]=[N-]
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)