missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Azido-N-(tert-butoxycarbonyl)-L-proline 98.0+%, TCI America™

Catalog No. A312950MG
Change view
Click to view available options
Quantity:
50 mg
200 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A312950MG 50 mg
A3129200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A312950MG Supplier TCI America Supplier No. A312950MG

Chemical Identifiers

CAS 132622-65-2
Molecular Formula C10H16N4O4
Molecular Weight (g/mol) 256.262
MDL Number MFCD04115791
InChI Key JZEOWBJXFSZTJU-BQBZGAKWSA-N
Synonym cis-4-Azido-N-Boc-L-proline, (2S,4S)-4-Azido-1-(tert-butoxycarbonyl)-2-pyrrolidinecarboxylic Acid, (2S,4S)-4-Azido-1-Boc-2-pyrrolidinecarboxylic Acid
PubChem CID 12094910
IUPAC Name (2S,4S)-4-azido-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC(CC1C(=O)O)N=[N+]=[N-]

Specifications

Melting Point 71°C
Color White-Yellow
Quantity 50 mg
Formula Weight 256.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material cis-4-Azido-N-(tert-butoxycarbonyl)-L-proline
TSCA No
Recommended Storage Refrigerator