accessibility menu, dialog, popup

UserName

NNOH

CAS RN 132213-07-1

IUPAC Name: {imidazo[1,2-a]pyridin-6-yl}methanol
Synonyms: imidazo[1,2-a]pyridin-6-ylmethanol
Molecular Weight (g/mol): 148.17
Molecular Formula: C8H8N2O
InChi Key: IEOADZOTWIMSMC-UHFFFAOYSA-N
SMILES: OCC1=CN2C=CN=C2C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)