missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridin-6-ylmethanol, 97%, Thermo Scientific™

Catalog No. p-4510487
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC33109CB 250mg
CC33109DA 1g
CC33109DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC33109CB Supplier Thermo Scientific Chemicals Supplier No. CC33109CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 132213-07-1
Molecular Formula C8H8N2O
Molecular Weight (g/mol) 148.165
InChI Key IEOADZOTWIMSMC-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridin-6-ylmethanol, 6-hydroxymethyl imidazo 1,2-a pyridine, imidazo 1,2-a pyridine-6-methanol, imidazo 1,2-a pyridin-6-yl-methanol, imidazo 1,2-a pyridin-6-yl methanol, acmc-209zmz, imidazo 1,2-a pyridin-6yl methanol, 6-hydroxymethyl imidazo 1,2-a pyridin, 4-hydroimidazo 1,2-a pyridin-6-ylmethan-1-ol
PubChem CID 7060530
IUPAC Name imidazo[1,2-a]pyridin-6-ylmethanol
SMILES C1=CC2=NC=CN2C=C1CO

Specifications

Assay Percent Range 97%
Quantity 250mg
Formula Weight 148.16
Percent Purity 97%
Chemical Name or Material Imidazo[1, 2-a]pyridin-6-ylmethanol