accessibility menu, dialog, popup

UserName

CAS RN 132178-78-0

IUPAC Name: 1,5-dibromonaphthalene-2,6-diol
Synonyms: 1,5-dibromonaphthalene-2,6-diol
Molecular Weight (g/mol): 317.96
Molecular Formula: C10H6Br2O2
InChi Key: COJNHIANORGBGY-UHFFFAOYSA-N
SMILES: OC1=CC=C2C(Br)=C(O)C=CC2=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results