accessibility menu, dialog, popup

UserName

CH3H3CSiClCH3CH3CH3H3C

CAS RN 13154-24-0

IUPAC Name: chlorotris(propan-2-yl)silane
Synonyms: chlorotriisopropylsilane
Molecular Weight (g/mol): 192.8
Molecular Formula: C9H21ClSi
InChi Key: KQIADDMXRMTWHZ-UHFFFAOYSA-N
SMILES: CC(C)[Si](Cl)(C(C)C)C(C)C
Percent Purity 
  • (3)
  • (3)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (3)
  • (9)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (9)