missing translation for 'onlineSavingsMsg'
Learn More

Triisopropylsilyl Chloride 95.0+%, TCI America™

Catalog No. T10785ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10785ML 5 mL
T107825ML 25 mL
T1078250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T10785ML Supplier TCI America Supplier No. T10785ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13154-24-0
Molecular Formula C9H21ClSi
Molecular Weight (g/mol) 192.80
MDL Number MFCD00009656
InChI Key KQIADDMXRMTWHZ-UHFFFAOYSA-N
Synonym triisopropylsilyl chloride, chlorotriisopropylsilane, triisopropylchlorosilane, chlorotris propan-2-yl silane, chloro triisopropyl silane, silane, chlorotris 1-methylethyl, tipscl, triisopropylsilylchloride, triisopropyl chlorosilane, triisopropylchlorosilane, redistilled
PubChem CID 139400
IUPAC Name chlorotris(propan-2-yl)silane
SMILES CC(C)[Si](Cl)(C(C)C)C(C)C

Specifications

Color Yellow
Boiling Point 96°C
Quantity 5 mL
UN Number 2987
Formula Weight 192.80
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Triisopropylsilyl Chloride
EINECSNumber (2)-2041
TSCA No