missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorotriisopropylsilane, 97+%
Description
Chlorotriisopropylsilane is used as a silylating agent in neucleotide synthesis. It is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals and in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chlorotriisopropylsilane is used as a silylating agent in neucleotide synthesis. It is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals and in organic synthesis.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents, strong bases, alcohols and amines.
Chlorotriisopropylsilane is used as a silylating agent in neucleotide synthesis. It is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals and in organic synthesis.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents, strong bases, alcohols and amines.
Specifications
Specifications
| Boiling Point | 200°C |
| Quantity | 250 g |
| Flash Point | 62°C (144°F) |
| Odor | Acrid |
| Refractive Index | 1.4535 |
| Sensitivity | Moisture sensitive |
| Density | 0.908 |
| UN Number | UN2987 |
| Beilstein | 1737446 |
| Solubility Information | Miscible with water. |
| Show More |
Safety and Handling
| Hazard Category | H227-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318-H227 Causes severe skin burns and eye damage. Causes serious eye damage. Combustible liquid. |
| Precautionary Statement | P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only