accessibility menu, dialog, popup

UserName

H3COOHOO

CAS RN 13110-96-8

IUPAC Name: 5-acetyl-2-hydroxybenzoate
Synonyms: 5-acetyl-2-hydroxybenzoate
Molecular Weight (g/mol): 179.15
Molecular Formula: C9H7O4
InChi Key: NZRDKNBIPVLNHA-UHFFFAOYSA-M
SMILES: CC(=O)C1=CC=C(O)C(=C1)C([O-])=O
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)