missing translation for 'onlineSavingsMsg'
Learn More

5-Acetylsalicylic acid, 98+%

Catalog No. AAA1166322
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1166322 100 g
AAA1166306 5 g
AAA1166314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1166322 Supplier Thermo Scientific Chemicals Supplier No. A1166322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13110-96-8 | C9H8O4 | 180.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13110-96-8
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00013978
InChI Key NZRDKNBIPVLNHA-UHFFFAOYSA-N
PubChem CID 83151
IUPAC Name 5-acetyl-2-hydroxybenzoic acid
SMILES CC(=O)C1=CC(=C(C=C1)O)C(=O)O

Specifications

Melting Point 214°C to 216°C
Flash Point 250°C (482°F)
Quantity 100 g
Beilstein 2096968
Sensitivity Moisture sensitive
Synonym 5-acetylsalicylic acid, 3-acetyl-6-hydroxybenzoic acid, 5-acetyl-2-hydroxy-benzoic acid, benzoic acid, 5-acetyl-2-hydroxy, 5-acetyl salicylic acid, acmc-1boxk, 5-acetylsalicylicacid, 5-acetyl-2-hydroxybenzoic acid, benzoic acid,5-acetyl-2-hydroxy
Formula Weight 180.15
Percent Purity ≥98%
Chemical Name or Material 5-Acetylsalicylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-037-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only