missing translation for 'onlineSavingsMsg'
Learn More

5-Acetylsalicylic Acid 98.0+%, TCI America™

Catalog No. A23785G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A23785G Supplier TCI America Supplier No. A23785G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13110-96-8
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00013978
InChI Key NZRDKNBIPVLNHA-UHFFFAOYSA-N
Synonym 5-acetylsalicylic acid, 3-acetyl-6-hydroxybenzoic acid, 5-acetyl-2-hydroxy-benzoic acid, benzoic acid, 5-acetyl-2-hydroxy, 5-acetyl salicylic acid, acmc-1boxk, 5-acetylsalicylicacid, 5-acetyl-2-hydroxybenzoic acid, benzoic acid,5-acetyl-2-hydroxy
PubChem CID 83151
IUPAC Name 5-acetyl-2-hydroxybenzoic acid
SMILES CC(=O)C1=CC(=C(C=C1)O)C(=O)O

Specifications

Melting Point 218°C
Color White-Yellow
Quantity 5 g
Formula Weight 180.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Acetylsalicylic Acid