CAS RN 13093-65-7
IUPAC Name:
(2S)-2-hydroxy-2-(4-hydroxyphenyl)acetonitrile
Synonyms:
(S)-4-hydroxymandelonitrile
Molecular Weight (g/mol):
149.15
Molecular Formula:
C8H7NO2
InChi Key:
HOOOPXDSCKBLFG-MRVPVSSYSA-N
SMILES:
O[C@H](C#N)C1=CC=C(O)C=C1
1
–
1
of
1
results
1
DL-4-Hydroxymandelonitrile 98.0+%, TCI America™
CAS: 13093-65-7 Molecular Formula: C8H7NO2 Molecular Weight (g/mol): 149.15 MDL Number: MFCD00056685 InChI Key: HOOOPXDSCKBLFG-MRVPVSSYSA-N Synonym: DL-alpha,4-Dihydroxyphenylacetonitrile PubChem CID: 166768 ChEBI: CHEBI:18115 IUPAC Name: (2S)-2-hydroxy-2-(4-hydroxyphenyl)acetonitrile SMILES: O[C@H](C#N)C1=CC=C(O)C=C1