missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(Iii) Oxide, purified, Solstice
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Physical Form | Crystalline Powder |
| Quantity | 250 g |
| UN Number | NONH for all modes of transport |
| Formula Weight | 159.69g/mol |
| Percent Purity | ≥95% |
| Grade | Purified |
| Chemical Name or Material | Iron(Iii) Oxide |
Safety and Handling
| EINECSNumber | 215-168-2 |
| RTECSNumber | NO7400000 |