missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermit(R), Granular, Spectrum™ Chemical
Specifications
Specifications
| Chemical Name or Material | Thermit(R), Granular |
| CAS | 1309-37-1 |
| Physical Form | Solid |
| Quantity | 25 g |
| Grade | Ungraded |
| Packaging | Poly Pail |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.