accessibility menu, dialog, popup

UserName

CAS RN 13057-72-2

IUPAC Name: 7-hydroxy-3-phenyl-4H-chromen-4-one
Synonyms: 7-hydroxyisoflavone
Molecular Weight (g/mol): 238.24
Molecular Formula: C15H10O3
InChi Key: WMKOZARWBMFKAS-UHFFFAOYSA-N
SMILES: OC1=CC=C2C(OC=C(C2=O)C2=CC=CC=C2)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results