missing translation for 'onlineSavingsMsg'
Learn More

7-Hydroxyisoflavone 98.0+%, TCI America™

Catalog No. H15791G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H15791G Supplier TCI America Supplier No. H15791G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 13057-72-2
Melting Point 216°C
Color White-Yellow
Molecular Formula C15H10O3
Quantity 1 g
MDL Number MFCD00017701
Synonym 7-Hydroxy-3-phenylchromone
InChI Key WMKOZARWBMFKAS-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=COC3=C(C2=O)C=CC(=C3)O
IUPAC Name 7-hydroxy-3-phenylchromen-4-one
Molecular Weight (g/mol) 238.242
PubChem CID 5376891
ChEBI CHEBI:12256
Formula Weight 238.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 7-Hydroxyisoflavone
Show More Show Less
RTECSNumber DJ3100255
TSCA No