accessibility menu, dialog, popup

UserName

CAS RN 13048-33-4

IUPAC Name: 6-(prop-2-enoyloxy)hexyl prop-2-enoate
Synonyms: 1,6-hexanediol diacrylate
Molecular Weight (g/mol): 226.27
Molecular Formula: C12H18O4
InChi Key: FIHBHSQYSYVZQE-UHFFFAOYSA-N
SMILES: C=CC(=O)OCCCCCCOC(=O)C=C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)