missing translation for 'onlineSavingsMsg'
Learn More

1,6-Bis(acryloyloxy)hexane (stabilized with MEHQ) 85.0+%, TCI America™

Catalog No. B2936500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2936500G 500 g
B293625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2936500G Supplier TCI America Supplier No. B2936500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13048-33-4
Molecular Formula C12H18O4
Molecular Weight (g/mol) 226.27
MDL Number MFCD00008631
InChI Key FIHBHSQYSYVZQE-UHFFFAOYSA-N
Synonym 1,6-hexanediol diacrylate, hexamethylene diacrylate, kayarad hdda, hexane-1,6-diyl diacrylate, 1,6-hexamethylene diacrylate, hexamethylene glycol diacrylate, sartomer 238, hdoda, viscoat 230, hexamethylene acrylate
PubChem CID 25644
IUPAC Name 6-(prop-2-enoyloxy)hexyl prop-2-enoate
SMILES C=CC(=O)OCCCCCCOC(=O)C=C

Specifications

Color Yellow
Boiling Point 165°C
Quantity 500 g
UN Number 3082
Formula Weight 226.27
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,6-Bis(acryloyloxy)hexane (stabilized with MEHQ)
EINECSNumber (2)-1007&(2)-0958
RTECSNumber AT1430000
TSCA Yes