missing translation for 'onlineSavingsMsg'
Learn More

Boron oxide, Puratronic™, (H{2}O 200ppm), 99.999% (metals basis), Thermo Scientific Chemicals

Catalog No. AA11160QE
Change view
Click to view available options
Quantity:
1 cyl
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11160QE 1 cyl
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA11160QE Supplier Thermo Scientific Chemicals Supplier No. 011160QE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1303-86-2 | Molecular Formula: B2O3 | Molecular Weight: 69.62 g/mol

In the electronics industry as a liquid encapsulent for Czochralski-type growth of III-V semiconductor single crystals (e.g., GaAs, GaP, InP).Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in the production of boron halides, esters, carbide, nitride and metallic borides. It has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants, heatproof glasswares and dopants in semiconductor materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In the electronics industry as a liquid encapsulent for Czochralski-type growth of III-V semiconductor single crystals (e.g., GaAs, GaP, InP)

Chemical Identifiers

CAS 1303-86-2
Molecular Formula B2O3
Molecular Weight (g/mol) 69.62
MDL Number MFCD00011315
InChI Key JKWMSGQKBLHBQQ-UHFFFAOYSA-N
Synonym boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
PubChem CID 518682
ChEBI CHEBI:30163
IUPAC Name oxo(oxoboranyloxy)borane
SMILES O=BOB=O

Specifications

Boiling Point 1860°C
Melting Point ∼450°C
Color White
Physical Form Cylinder
Quantity 1 cyl
Odor Odorless
Sensitivity Hygroscopic
Density 2.46 g/mL (±)0.01
Merck Index 14,1337
Solubility Information Slightly soluble in water and methanol.
Formula Weight 69.62
Percent Purity 99.999%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Boron oxide, H2O 200ppm, Cylinder, 5.0cm dia.
Show More Show Less
Hazard Category H360FD
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 215-125-8
RTECSNumber ED7900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only