missing translation for 'onlineSavingsMsg'
Learn More

Boric Anhydride, Purified, 97%, Spectrum™ Chemical

Catalog No. 18609190
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609190 50 kg Poly Bottle
18609187 125 g Poly Bottle
18609189 500 g Poly Bottle
18609186 2.5 kg Poly Bottle
18609188 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18609190 Supplier Spectrum Chemical Mfg Cor Supplier No. B113550KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1303-86-2

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 50 kg
Percent Purity 97%
Grade Purified
Packaging Poly Bottle
Chemical Name or Material Boric Anhydride

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.