accessibility menu, dialog, popup

UserName

OSOOFFFI

CAS RN 129112-26-1

IUPAC Name: 2-iodophenyl trifluoromethanesulfonate
Synonyms: 2-iodophenyl trifluoromethanesulfonate
Molecular Weight (g/mol): 352.07
Molecular Formula: C7H4F3IO3S
InChi Key: AXIDYNSWXPNBLG-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)OC1=CC=CC=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results