CAS RN 129-06-6
CAS RN 129-06-6
IUPAC Name:
sodium 2,4-dioxo-3-(3-oxo-1-phenylbutyl)-3,4-dihydro-2H-1-benzopyran-3-ide
Synonyms:
warfarin sodium
Molecular Weight (g/mol):
330.32
Molecular Formula:
C19H15NaO4
InChi Key:
PAIQCHHPKBVDOM-UHFFFAOYNA-N
SMILES:
[Na+].CC(=O)CC([C-]1C(=O)OC2=CC=CC=C2C1=O)C1=CC=CC=C1
1
–
2
of
2
results
2
Warfarin Sodium (contains Isopropyl Alcohol) 98.0+%, TCI America™
CAS: 129-06-6 Molecular Formula: C19H18NaO4 Molecular Weight (g/mol): 333.339 MDL Number: MFCD00083223 InChI Key: MBNHFFMQYKZCLK-UHFFFAOYSA-N Synonym: 3-(alpha-Acetonylbenzyl)-4-hydroxycoumarin Sodiuim Salt, 4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin Sodium Salt PubChem CID: 131664125 IUPAC Name: 4-hydroxy-3-(3-oxo-1-phenylbutyl)chromen-2-one;molecular hydrogen;sodium SMILES: [HH].CC(=O)CC(C1=CC=CC=C1)C2=C(C3=CC=CC=C3OC2=O)O.[Na]