missing translation for 'onlineSavingsMsg'
Learn More

Warfarin Sodium Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019445
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019445 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019445 Supplier Merck Emd Millipore Supplier No. PHR14351G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards. Warfarin Sodium is an anticoagulant rodenticide, commonly used for the control of rodents such as rats, mice, etc. in surroundings, commercial and industrial sites.

Specifications

CAS 129-06-6
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C19H15O4 Na
MDL Number MFCD00083223
Synonym 3-(alpha-Acetonylbenzyl)-4-hydroxycoumarin sodium salt; 4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin
Type Certified Reference Material
Quantity 1 g
Formula Weight 330.31
Grade Certified Reference Material