accessibility menu, dialog, popup

UserName

CAS RN 127-69-5

IUPAC Name: 4-amino-N-(3,4-dimethyl-1,2-oxazol-5-yl)benzene-1-sulfonamide
Synonyms: ganda soxazole astrazolo accuzole gantrisona sulfisoxazole (substance) dorsulfan warthausen neazolin V sul roche brand of sulfisoxazole diolamine
Molecular Weight (g/mol): 267.3
Molecular Formula: C11H13N3O3S
InChi Key: NHUHCSRWZMLRLA-UHFFFAOYSA-N
SMILES: CC1=NOC(NS(=O)(=O)C2=CC=C(N)C=C2)=C1C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)