accessibility menu, dialog, popup

UserName

HOBOHN

CAS RN 126747-14-6

IUPAC Name: (4-cyanophenyl)boronic acid
Synonyms: 4-cyanophenylboronic acid
Molecular Weight (g/mol): 146.94
Molecular Formula: C7H6BNO2
InChi Key: CEBAHYWORUOILU-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)C#N
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)