missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C17785G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17785G 5 g
C177825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17785G Supplier TCI America Supplier No. C17785G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 126747-14-6
Molecular Formula C7H6BNO2
Molecular Weight (g/mol) 146.94
MDL Number MFCD01318968
InChI Key CEBAHYWORUOILU-UHFFFAOYSA-N
Synonym 4-cyanophenyl boronic acid, 4-cyanobenzeneboronic acid, 4-dihydroxyboranyl benzonitrile, 4-cyano-phenyl-boronic acid, 4-boronobenzonitrile
PubChem CID 2734326
IUPAC Name (4-cyanophenyl)boronic acid
SMILES OB(O)C1=CC=C(C=C1)C#N

Specifications

Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 146.94
Physical Form Crystalline Powder