missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzeneboronic acid, 98%

Catalog No. AAL1800706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1800706 5 g
AAL1800703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1800706 Supplier Thermo Scientific Chemicals Supplier No. L1800706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 126747-14-6 | C7H6BNO2 | 146.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 126747-14-6
Molecular Formula C7H6BNO2
Molecular Weight (g/mol) 146.94
MDL Number MFCD01318968
InChI Key CEBAHYWORUOILU-UHFFFAOYSA-N
Synonym 4-cyanophenyl boronic acid, 4-cyanobenzeneboronic acid, 4-dihydroxyboranyl benzonitrile, 4-cyano-phenyl-boronic acid, 4-boronobenzonitrile, p-cyanophenylboronic acid, 4-cyanobenzene boronic acid, boronic acid, 4-cyanophenyl, 4-cyano-phenyl boronic acid
PubChem CID 2734326
IUPAC Name (4-cyanophenyl)boronic acid
SMILES OB(O)C1=CC=C(C=C1)C#N

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 6593772
Formula Weight 146.94
Percent Purity 98%
Chemical Name or Material 4-Cyanobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only