CAS RN 126690-96-8
IUPAC Name:
1-bromotetracyclo[6.6.2.0²,⁷.0⁹,¹⁴]hexadeca-2,4,6,9,11,13,15-heptaene
Synonyms:
1-bromotetracyclo[6.6.2.0²,⁷.0⁹,¹⁴]hexadeca-2,4,6,9,11,13,15-heptaene
Molecular Weight (g/mol):
283.17
Molecular Formula:
C16H11Br
InChi Key:
JNPBTNVWRDISGS-UHFFFAOYSA-N
SMILES:
BrC12C=CC(C3=CC=CC=C13)C1=CC=CC=C21
1
–
1
of
1
results
1
9-Bromo-9,10-dihydro-9,10-ethenoanthracene 98.0+%, TCI America™
CAS: 126690-96-8 Molecular Formula: C16H11Br Molecular Weight (g/mol): 283.168 InChI Key: JNPBTNVWRDISGS-UHFFFAOYSA-N Synonym: 9-Bromodibenzobarrelene PubChem CID: 91972153 SMILES: C1=CC=C2C(=C1)C3C=CC2(C4=CC=CC=C34)Br