accessibility menu, dialog, popup

UserName

OHOHOHNOO

CAS RN 126-11-4

IUPAC Name: 2-(hydroxymethyl)-2-nitropropane-1,3-diol
Synonyms: tris-hydroxymethylnitromethane
Molecular Weight (g/mol): 151.12
Molecular Formula: C4H9NO5
InChi Key: OLQJQHSAWMFDJE-UHFFFAOYSA-N
SMILES: OCC(CO)(CO)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)