missing translation for 'onlineSavingsMsg'
Learn More

Tris(hydroxymethyl)nitromethane, 95%

Catalog No. AAL1334936
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1334936 500 g
AAL1334922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1334936 Supplier Thermo Scientific Chemicals Supplier No. L1334936
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 126-11-4 | C4H9NO5 | 151.12 g/mol

Tris(hydroxymethyl)nitromethane, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(hydroxymethyl)nitromethane, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as a primary and secondary intermediate.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, bases. Stable under recommended storage conditions.

Chemical Identifiers

CAS 126-11-4
Molecular Formula C4H9NO5
Molecular Weight (g/mol) 151.12
MDL Number MFCD00007395
InChI Key OLQJQHSAWMFDJE-UHFFFAOYSA-N
Synonym tris hydroxymethyl nitromethane, 2-hydroxymethyl-2-nitropropane-1,3-diol, 2-hydroxymethyl-2-nitro-1,3-propanediol, cimcool wafers, tris nitro, trimethylolnitromethane, isobutylglycerol, nitro, nitroisobutylglycerol, 1,3-propanediol, 2-hydroxymethyl-2-nitro, methane, trimethylolnitro
PubChem CID 31337
IUPAC Name 2-(hydroxymethyl)-2-nitropropane-1,3-diol
SMILES OCC(CO)(CO)[N+]([O-])=O

Specifications

Melting Point ∽160°C (decomposition)
Linear Formula (HOCH2)3CNO2
Quantity 500 g
Beilstein 742427
Merck Index 14,9753
Solubility Information It is soluble in water.
Formula Weight 151.12
Percent Purity 95%
Chemical Name or Material Tris(hydroxymethyl)nitromethane
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
EINECSNumber 204-769-5
RTECSNumber TY7350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only