accessibility menu, dialog, popup

UserName

CAS RN 125971-95-1

H3CCH3NOOH3CCH3OOH3CH3CCH3FOHN

CAS RN 125971-95-1

IUPAC Name: tert-butyl 2-(6-{2-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-(propan-2-yl)-1H-pyrrol-1-yl]ethyl}-2,2-dimethyl-1,3-dioxan-4-yl)acetate
Synonyms: tert-butyl 2-(6-{2-[2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)pyrrol-1-yl]ethyl}-2,2-dimethyl-1,3-dioxan-4-yl)acetate
Molecular Weight (g/mol): 654.82
Molecular Formula: C40H47FN2O5
InChi Key: NPPZOMYSGNZDKY-UHFFFAOYNA-N
SMILES: CC(C)C1=C(C(=O)NC2=CC=CC=C2)C(=C(N1CCC1CC(CC(=O)OC(C)(C)C)OC(C)(C)O1)C1=CC=C(F)C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results