missing translation for 'onlineSavingsMsg'
Learn More

Atorvastatin Related Compound I Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012850
Change view
Click to view available options
Quantity:
30 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012850 30 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012850 Supplier Merck Emd Millipore Supplier No. PHR187430MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This material is an impurity of atorvastatin. Atorvastatin is a synthetic HMG-CoA reductase inhibitor which lowers plasma cholesterol levels by inhibiting endogenous cholesterol synthesis.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 125971-95-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C40H47 FN2O5
MDL Number MFCD04039904
Melting Point 144°C to 148°C
Synonym tert-Butyl (4 R,6 R)-6-[2-[2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1-pyrrolyl]ethyl]-2,2-dimethyl-1,3-dioxane-4-acetate; tert-Butyl 2-((4 R,6 R)-6-{2-[2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1 H-pyrrol-1-yl]ethyl}-2,2-dimethyl-1,3-dioxan-4-yl)acetate; (4 R,6 R)-6-[2-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1 H-pyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxane-4-acetic acid 1,1-dimethylethyl ester; tert-Butyl 2-[(4 R,6 R)-6-[2-[2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1 H-pyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxan-4-yl]acetate
Type Certified Reference Material
Quantity 30 mg
Formula Weight 654.81
Grade Certified Reference Material