accessibility menu, dialog, popup

UserName

CH3H3CCH3OO(R)(R)NOH3CCH3O

CAS RN 125971-94-0

IUPAC Name: tert-butyl 2-[(4R,6R)-6-(cyanomethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate
Synonyms: tert-butyl 2-[(4R,6R)-6-(cyanomethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate
Molecular Weight (g/mol): 269.34
Molecular Formula: C14H23NO4
InChi Key: DPNRMEJBKMQHMC-GHMZBOCLSA-N
SMILES: CC(C)(C)OC(=O)C[C@H]1C[C@@H](CC#N)OC(C)(C)O1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)